Skip to Content
Merck
Rubidium sulfate 99.8% trace metals basis

Rubidium sulfate

Rubidium sulfate 99.8% trace metals basis
Synonym(s):
Sulfuric acid, dirubidium salt
Linear Formula:
Rb2SO4
CAS Number:
Molecular Weight:
267.00
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing