Skip to Content
Merck
Rink amide resin 50-90 mesh, extent of labeling: 0.8-1.0 mmol/g N loading

Rink amide resin

Rink amide resin 50-90 mesh, extent of labeling: 0.8-1.0 mmol/g N loading
Synonym(s):
Rink acid-labil (hydroxymethyl)polystyrene

Products

Product Category
Brand
Functional Group
Application
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing