
(−)-Riboflavin (Vitamin B2) solution

Synonym(s):
(−)-Riboflavin, Lactoflavin, Vitamin B2, Vitamin G
Empirical Formula (Hill Notation):
C17H20N4O6
CAS Number:
Molecular Weight:
376.36
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing