Skip to Content
Merck
(R)-γ-Valerolactone ≥97.5% (GC)

(R)-γ-Valerolactone

(R)-γ-Valerolactone ≥97.5% (GC)
Synonym(s):
(5R)-4-Hydroxypentanoic acid lactone, (5R)-5-Methyloxolan-2-one, (R)-γ-Methyl-γ-butyrolactone, (R)-5-Methyl-2-oxotetrahydrofuran, (R)-Dihydro-5-methyl-2(3H)-furanone
Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing