Skip to Content
Merck
(R)-(−)-Glycidyl benzyl ether 99%

(R)-(−)-Glycidyl benzyl ether

(R)-(−)-Glycidyl benzyl ether 99%
Synonym(s):
(R)-(−)-2-(Benzyloxymethyl)oxirane
Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing