Skip to Content
Merck
(R)-(+)-N,N-Dimethyl-1-phenylethylamine 97%

(R)-(+)-N,N-Dimethyl-1-phenylethylamine

(R)-(+)-N,N-Dimethyl-1-phenylethylamine 97%
Synonym(s):
(R)-(+)-N,N,α-Trimethylbenzylamine
Linear Formula:
C6H5CH(CH3)N(CH3)2
CAS Number:
Molecular Weight:
149.23
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing