Skip to Content
Merck
(R)-(+)-N-Benzyl-α-methylbenzylamine 98%

(R)-(+)-N-Benzyl-α-methylbenzylamine

(R)-(+)-N-Benzyl-α-methylbenzylamine 98%
Synonym(s):
(R)-(+)-N-(1-Phenylethyl)benzylamine, (R)-(+)-N-Benzyl-1-phenylethylamine
Linear Formula:
C6H5CH(CH3)NHCH2C6H5
CAS Number:
Molecular Weight:
211.30

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing