Skip to Content
Merck
(R)-(+)-N,α-Dimethylbenzylamine 98%

(R)-(+)-N,α-Dimethylbenzylamine

(R)-(+)-N,α-Dimethylbenzylamine 98%
Synonym(s):
(R)-(+)-N-Methyl-1-phenylethylamine
Linear Formula:
C6H5CH(CH3)NHCH3
CAS Number:
Molecular Weight:
135.21

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing