Skip to Content
Merck
(R)-(+)-4-Methyl-4-(trichloromethyl)-2-oxetanone 98%

(R)-(+)-4-Methyl-4-(trichloromethyl)-2-oxetanone

(R)-(+)-4-Methyl-4-(trichloromethyl)-2-oxetanone 98%
Synonym(s):
(R)-(+)-3-Hydroxy-3-methyl-4,4,4-trichlorobutyric acid β-lactone
Empirical Formula (Hill Notation):
C5H5Cl3O2
CAS Number:
Molecular Weight:
203.45

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing