
(R)-(+)-3-Chlorostyrene oxide

Synonym(s):
(R)-(+)-(3-Chlorophenyl)oxirane
Empirical Formula (Hill Notation):
C8H7ClO
CAS Number:
Molecular Weight:
154.59
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing