
(R)-(+)-2-(Methoxymethoxy)-1-propanol

Synonym(s):
MOM protected (R)-1,2-Propanediol
Empirical Formula (Hill Notation):
C5H12O3
CAS Number:
Molecular Weight:
120.15
Products
Product Category
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing