Skip to Content
Merck
(R)-(+)-1-Phenyl-1-propanol 99%

(R)-(+)-1-Phenyl-1-propanol

(R)-(+)-1-Phenyl-1-propanol 99%
Synonym(s):
(R)-(+)-α-Ethylbenzyl alcohol
Linear Formula:
C2H5CH(C6H5)OH
CAS Number:
Molecular Weight:
136.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing