Skip to Content
Merck
(R)-(+)-1-(2-Naphthyl)ethanol 98%

(R)-(+)-1-(2-Naphthyl)ethanol

(R)-(+)-1-(2-Naphthyl)ethanol 98%
Synonym(s):
(R)-(+)-α-Methyl-2-naphthalenemethanol
Linear Formula:
C10H7CH(CH3)OH
CAS Number:
Molecular Weight:
172.22

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing