Skip to Content
Merck
(R)-(−)-Citronellyl bromide 95%

(R)-(−)-Citronellyl bromide

(R)-(−)-Citronellyl bromide 95%
Synonym(s):
8-Bromo-2,6-dimethyl-2-octene
Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2Br
CAS Number:
Molecular Weight:
219.16

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing