Skip to Content
Merck
(R)-(−)-2-Methyl-2,4-pentanediol 99%

(R)-(−)-2-Methyl-2,4-pentanediol

(R)-(−)-2-Methyl-2,4-pentanediol 99%
Synonym(s):
(−)-Hexylene glycol
Linear Formula:
CH3CH(OH)CH2C(CH3)2OH
CAS Number:
Molecular Weight:
118.17

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing