
(R)-(−)-1-Methyl-3-pyrrolidinol

Synonym(s):
(3R)-1-Methylpyrrolidin-3-ol, (R)-(−)-3-Hydroxy-N-methylpyrrolidine, (R)-N-Methyl-3-pyrrolidinol
Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing