Propiconazole-(phenyl-d3)
Synonym(s):
1-{[2-(2,4-Dichlorophenyl-d3)-4-propyl-1,3-dioxolan-2-yl]methyl}-1H-1,2,4-triazole, Propiconazole-d3
Empirical Formula (Hill Notation):
C15D3H14Cl2N3O2
Molecular Weight:
345.24
Products
Product Category
Brand
Formula Weight
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing