Skip to Content
Merck
Potassium (4-cyanophenyl)trifluoroborate 97%

Potassium (4-cyanophenyl)trifluoroborate

Potassium (4-cyanophenyl)trifluoroborate 97%
Empirical Formula (Hill Notation):
C7H4BF3KN
CAS Number:
Molecular Weight:
209.02

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing