
Poly(N-isopropylacrylamide), maleimide terminated

Synonym(s):
P(NIPAM)n-NH-maleimide, Polyacrylamide, functionalized polyNIPAM, functionalized polyacrylamide, polyNIPAM
Linear Formula:
(C6H11NO)n SCH2CH2NH(CO)CH2CH2CH2(C4H2NO2)
Products
Brand
Physical Form
Polymer Composition
Polymer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing