
Poly(L-lactide) N-2-hydroxyethylmaleimide terminated

Synonym(s):
PLA maleimide, PLLA MI terminated
Linear Formula:
C6H7NO3(C3H4O2)n
Products
Brand
Melting Point (°C)
Physical Form
Optical Isomer
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing