Poly(L-lactide), 2-bromoisobutyryl terminated
Synonym(s):
ATRP Macromonomer, ATRP terminated PLLA, PLA
Linear Formula:
H(C3H4O2)nC6H10BrO3
Products
Brand
Melting Point (°C)
Physical Form
Optical Isomer
Polymer Composition
Polymer Type
Showing 1-2 of 2