
Poly(ethylene glycol) bis(2-bromoisobutyrate)

Synonym(s):
PEG ATRP initiator, PEG BIBB
Linear Formula:
C4H6O2Br(C2H4O)nC4H6OBr
Products
Brand
Melting Point (°C)
Physical Form
Polymer Composition
Polymer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing