
Poly(ethylene glycol), 16 hydroxyl dendron, generation 3

Synonym(s):
Dendron functionalized PEG, generation 3
Products
Product Category
Brand
Melting Point (°C)
Physical Form
Generation
Functional Group
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing