Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(3,4-ethylenedioxythiophene), bis-poly(ethyleneglycol), lauryl terminated
Synonym(s):
C
12
-PEG-
block
-PEDOT-
block
-PEG-C
12
, PEDOT:PEG
Products
Brand
Sigma-Aldrich (2)
Physical Form
Chemical Composition
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
736309
Poly(3,4-ethylenedioxythiophene), bis-poly(ethyleneglycol), lauryl terminated,
0.8 wt. % (dispersion in 1,2-dichlorobenzene), contains
p
-toluenesulfonate as dopant
Expand
View Pricing
736295
Poly(3,4-ethylenedioxythiophene), bis-poly(ethyleneglycol), lauryl terminated,
0.7 wt. % (dispersion in nitromethane), contains
p
-toluenesulfonate as dopant
Expand
View Pricing