Skip to Content
Merck
Plexcore® OC RG-1110 organic conductive ink

Plexcore® OC RG-1110 organic conductive ink

Plexcore® OC RG-1110 organic conductive ink
Synonym(s):
Plexcore® OC organic conductive ink

Products

Brand
pH Value
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing