Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PLA 3D printing filament
CAS Number:
9051-89-2
Products
Brand
Sigma-Aldrich (9)
Sustainability
Color
Feature
Diameter (mm)
Special Grade
Natural (2)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
3DXPLA007
PLA 3D printing filament,
natural, diam. 1.75 mm
Expand
View Pricing
3DXPLA011
PLA 3D printing filament,
red, diam. 1.75 mm
Expand
View Pricing
3DXPLA008
PLA 3D printing filament,
natural, diam. 2.85 mm
Expand
View Pricing
3DXPLA015
PLA 3D printing filament,
yellow, diam. 1.75 mm
Expand
View Pricing
3DXPLA006
PLA 3D printing filament,
green, diam. 2.85 mm
Expand
View Pricing
3DXPLA009
PLA 3D printing filament,
orange, diam. 1.75 mm
Expand
View Pricing
3DXPLA016
PLA 3D printing filament,
yellow, diam. 2.85 mm
Expand
View Pricing
3DXPLA012
PLA 3D printing filament,
red, diam. 2.85 mm
Expand
View Pricing
3DXPLA010
PLA 3D printing filament,
orange, diam. 2.85 mm
Expand
View Pricing