Skip to Content
Merck
Phenanthrene solution certified reference material, 5000 μg/mL in methanol

Phenanthrene solution

Phenanthrene solution certified reference material, 5000 μg/mL in methanol
CAS Number:

Products

Brand
Gene Alias
Product Line
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing