Skip to Content
Merck
Phenacyl 4-(bromomethyl)phenylacetate 95%

Phenacyl 4-(bromomethyl)phenylacetate

Phenacyl 4-(bromomethyl)phenylacetate 95%
Synonym(s):
4-(Bromomethyl)phenylacetic acid phenacyl ester
Empirical Formula (Hill Notation):
C17H15BrO3
CAS Number:
Molecular Weight:
347.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing