Skip to Content
Merck
Phe-Gly hydrate ≥98.0% (dried material)

Phe-Gly hydrate

Phe-Gly hydrate ≥98.0% (dried material)
Synonym(s):
L-Phenylalanyl-glycine hydrate
Linear Formula:
C6H5CH2CH(NH2)CONHCH2COOH · aq
CAS Number:
Molecular Weight:
222.24 (anhydrous basis)

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing