
Pentamethylenebis(magnesium bromide) solution

Synonym(s):
Pentane-1,5-di(magnesium bromide)
Linear Formula:
BrMg(CH2)5MgBr
CAS Number:
Molecular Weight:
278.55
Products
Brand
Formula Weight
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing