Skip to Content
Merck
Oxotremorine sesquifumarate salt ≥98% (HPLC), solid

Oxotremorine sesquifumarate salt

Oxotremorine sesquifumarate salt ≥98% (HPLC), solid
Synonym(s):
1-(4-[1-Pyrrolidinyl]-2-butynyl)-2-pyrrolidinone, 1-(4-[1-Pyrrolidinyl]2-butynyl)-2-pyrrolidinone
Empirical Formula (Hill Notation):
C12H18N2O · 1.5C4H4O4 · xH2O
CAS Number:
Molecular Weight:
380.39 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing