Skip to Content
Merck
Octyl α-D-glucopyranoside ≥98% (GC)

Octyl α-D-glucopyranoside

Octyl α-D-glucopyranoside ≥98% (GC)
Synonym(s):
n-Octyl α-glucoside
Empirical Formula (Hill Notation):
C14H28O6
CAS Number:
Molecular Weight:
292.37

Products

Product Category
Brand
Formula Weight
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing