Skip to Content
Merck
N-Z-Ethylenediamine hydrochloride 95%

N-Z-Ethylenediamine hydrochloride

N-Z-Ethylenediamine hydrochloride 95%
Synonym(s):
N-Z-1,2-Diaminoethane hydrochloride, Benzyl N-(2-aminoethyl)carbamate hydrochloride
Linear Formula:
C6H5CH2OCONHCH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
230.69

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing