Skip to Content
Merck
N-Z-Ethanolamine 98%

N-Z-Ethanolamine

N-Z-Ethanolamine 98%
Synonym(s):
2-(Z-Amino)ethanol, Benzyl N-(2-hydroxyethyl)carbamate
Linear Formula:
HOCH2CH2NHCO2CH2C6H5
CAS Number:
Molecular Weight:
195.22

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing