
N-Suberoylglycine

Synonym(s):
8-[(Carboxymethyl)amino]-8-oxooctanoic acid, N-Suberylglycine
Empirical Formula (Hill Notation):
C10H17NO5
CAS Number:
Molecular Weight:
231.25
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing