Skip to Content
Merck
N-sec-Butyl(trimethylsilyl)amine 98%

N-sec-Butyl(trimethylsilyl)amine

N-sec-Butyl(trimethylsilyl)amine 98%
Empirical Formula (Hill Notation):
C7H19NSi
CAS Number:
Molecular Weight:
145.32

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing