Norfluoxetine hydrochloride
Synonym(s):
(±)-γ-(4-Trifluoromethylphenoxy)benzenepropanamine hydrochloride
Empirical Formula (Hill Notation):
C16H16F3NO · HCl
CAS Number:
Molecular Weight:
331.76
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing