Skip to Content
Merck
N,N′-Ethylenebis(acrylamide) technical grade, ≥90% (TLC)

N,N′-Ethylenebis(acrylamide)

N,N′-Ethylenebis(acrylamide) technical grade, ≥90% (TLC)
Synonym(s):
N,N′-Dimethylenebis(acrylamide), N,N′-Bisacryloyl-1,2-diaminoethane
Linear Formula:
(H2C=CHCONHCH2-)2
CAS Number:
Molecular Weight:
168.19

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing