Skip to Content
Merck
N,N-Diethyl-2-(1-naphthyloxy)propanamide 97%

N,N-Diethyl-2-(1-naphthyloxy)propanamide

N,N-Diethyl-2-(1-naphthyloxy)propanamide 97%
Linear Formula:
C10H7OCH(CH3)CON(CH2CH3)2
CAS Number:
Molecular Weight:
271.35
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing