Skip to Content
Merck
N-Hydroxyurethane

N-Hydroxyurethane

N-Hydroxyurethane
Synonym(s):
N-Carbethoxyhydroxylamine, Ethyl N-hydroxycarbamate, Hydroxycarbamic acid ethyl ester, NSC 71045, NSC 83629
Linear Formula:
HONHCOOCH2CH3
CAS Number:
Molecular Weight:
105.09
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing