Nessler’s reagent
Synonym(s):
Potassium tetraiodomercurate – Potassium hydroxide solution
CAS Number:
Products
Brand
Physical Form
Analyte Suitability
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing