
Neopentyl glycol dimethylsulfate

Synonym(s):
2,2-Dimethyl-1,3-propanediol dimethylsulfate
Empirical Formula (Hill Notation):
C7H16O6S2
CAS Number:
Molecular Weight:
260.33
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing