Skip to Content
Merck
N-Benzoyl-L-tyrosine p-amidobenzoic acid sodium salt lyophilized, ≥98.0% (sum of enantiomers, HPLC)

N-Benzoyl-L-tyrosine p-amidobenzoic acid sodium salt

N-Benzoyl-L-tyrosine p-amidobenzoic acid sodium salt lyophilized, ≥98.0% (sum of enantiomers, HPLC)
Synonym(s):
N-Benzoyl-L-tyrosyl-4-aminobenzoic acid sodium salt, Bentiromide
Linear Formula:
C23H19N2O5Na
CAS Number:
Molecular Weight:
444.41
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1