Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nafion
™
perfluorinated resin solution
CAS Number:
31175-20-9
Products
Product Category
resins (4)
Brand
Sigma-Aldrich (4)
Sustainability
Boiling Point (°C)
Feature
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
527084
Nafion
™
perfluorinated resin solution,
5 wt. % in mixture of lower aliphatic alcohols and water, contains 45% water
Expand
View Pricing
510211
Nafion
™
perfluorinated resin solution,
5 wt. % in mixture of lower aliphatic alcohols and water, contains 45% water
Expand
View Pricing
663492
Nafion
™
perfluorinated resin solution,
20 wt. % in lower aliphatic alcohols and water, contains 34% water
Expand
View Pricing
527122
Nafion
™
perfluorinated resin solution,
20 wt. % in mixture of lower aliphatic alcohols and water, contains 34% water
Expand
View Pricing