N-Acetylneuraminyl-fucosyllacto-N-neo-tetraose from human milk
Synonym(s):
α-NeuNAc-(2→6)-β-Gal-(1→4)-β-GlcNAc-(1→3)-β-Gal-(1→4)-(α-Fuc-[1→3])-Glc, SFLNnT, Sialyl-fucosyllacto-N-neo-tetraose
Empirical Formula (Hill Notation):
C43H72N2O33
CAS Number:
Molecular Weight:
1145.03
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Tissue
Showing 1-1 of 1
Compare
Product Number
Description
Pricing