
N6-Cyclohexyladenosine

Synonym(s):
CHA
Empirical Formula (Hill Notation):
C16H23N5O4
CAS Number:
Molecular Weight:
349.38
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing