Skip to Content
Merck
N-(2-Fluorenyl)acetamide ≥98% (HPLC)

N-(2-Fluorenyl)acetamide

N-(2-Fluorenyl)acetamide ≥98% (HPLC)
Synonym(s):
2-AAF, 2-Acetamidofluorene, N-Acetyl-2-aminofluorene
Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing