Skip to Content
Merck
myo-Inositol 2-monophosphate bis(cyclohexylammonium) salt ~95%, powder

myo-Inositol 2-monophosphate bis(cyclohexylammonium) salt

myo-Inositol 2-monophosphate bis(cyclohexylammonium) salt ~95%, powder
Empirical Formula (Hill Notation):
C6H13O9P · 2C6H13N
CAS Number:
Molecular Weight:
458.48

Products

Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing