Skip to Content
Merck
Moupinamide ≥95% (LC/MS-ELSD)

Moupinamide

Moupinamide ≥95% (LC/MS-ELSD)
Synonym(s):
Alfrutamide
Empirical Formula (Hill Notation):
C18H19NO4
CAS Number:
Molecular Weight:
313.35

Products

Product Category
Brand
Formula Weight
Physical Form
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing