Skip to Content
Products
Cart
0
TH
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Monolayer graphene film
Products
Brand
Sigma-Aldrich (2)
Physical Form
Length (cm)
Length (in)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
773697
Monolayer graphene film,
1 cm x 1 cm on copper foil, avg. no. of layers, 1
Expand
View Pricing
745863
Monolayer graphene film,
1 in x 1 in on PET film, avg. no. of layers, 1
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
ไทย - TH
English - EN
Learn More